Please login to your online account to display your discounted pricing

Alfa Aesar™ Prepro-VIP/PHM (111-122)

For cellular and molecular biology applications

Specifications

Product Type Prepro-VIP/PHM (111-122)
Molecular Weight 1242.5
Storage Requirements -30° to -10°C
Format Solid
Sequence Val-Ser-Ser-Asn-Ile-Ser-Glu-Asp-Pro-Val-Pro-Val
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66910LB0

 
alfa aesar™
J66910LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66910MCR

 
alfa aesar™
J66910MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Prepro-VIP/PHM (111-122)
Molecular Weight 1242.5
Storage Requirements -30° to -10°C
Format Solid
Sequence Val-Ser-Ser-Asn-Ile-Ser-Glu-Asp-Pro-Val-Pro-Val
Common Name PHM-VIP Space Peptide
Regulatory Status RUO
Purification Method HPLC