Please login to your online account to display your discounted pricing

Alfa Aesar™ Prepro-VIP/PHM (111-122)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66910MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66910MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Storage Requirements -30° to -10°C
Format Solid
Sequence Val-Ser-Ser-Asn-Ile-Ser-Glu-Asp-Pro-Val-Pro-Val
Regulatory Status RUO
Common Name PHM-VIP Space Peptide
Product Type Prepro-VIP/PHM (111-122)
Molecular Weight 1242.5
Purification Method HPLC