missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Prepared Culture Media in Plates
Labeled with product name, expiration date and lot number.
Manufacturer: Fisher Science Education CM149P25
Description
- Sterile plates are ready for immediate use
- Supplied in disposable plastic Petri dishes, 100mm O.D. × 15mm deep
- Packed in clear, heat-sealed cellophane bags
Specifications
Plate | |
Potato Dextrose Agar culture media | |
Isolation, identification and cultivation of Yeasts and molds from dairy and food products | |
10/Pk. | |
15 mm | |
100 mm |
US | |
Sterile | |
Petri Dish | |
0.59 in. | |
3.93 in. |