Contec™ PREempt™ HLD5
PREempt™ HLD5 is a 2% AHP™ solution designed for instrument and device reprocessing of semi-critical and critical devices where rapid contact times and high-level disinfection are required
Supplier:  Contec™ 11505
Catalog No. 29-636-6288
Description
Description
  • High level disinfectant with 5 min. contact time against bacteria, viruses, fungi and mycobacteria
  • Disinfectant and sterilant for processing heat sensitive semi-critical and critical medical devices and instruments for which heat sterilization is not suitable
  • Contains hydrogen peroxide which converts to water and oxygen
  • Can be reused for 14 days after decanting
  • No special ventilation required
Specifications
Specifications
Disinfectant Solution
Non-sterile
2% AHP™
4 L