missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Precisionware™ High-Density Polyethylene Wide-Mouth Bottles

Ideal for packaging and storing of liquids and solids.

Manufacturer:  Bel-Art™ 106250005

 View more versions of this product

Catalog No. S95194



Description

Description

  • Rigid, leakproof design with easy-to-fill wide mouth
  • High-density polyethylene bottles with polypropylene closures
  • Feature recycling code molded into base, except S95194

Order Info

Pack of 12, except S95187, which comes in packs of 6

Specifications

Specifications

4.2 oz., 125 mL
4.2 oz.
HDPE
38 mm
Screw Cap
125 mL
Translucent
38
Polypropylene
48.2 mm
SDS
Documents
Product Certifications
Promotions

Promotions