Please login to your online account to display your discounted pricing

Thermo Scientific™ Precision Plant Growth Chambers

Implement uniform illumination with Thermo Scientific™ Precision Plant Growth Chambers. Designed for applications requiring precise temperature control and uniformity.

Specifications

Capacity 17.8 cu. ft. (504L)
Temperature Range (Metric) -10° to +50°C (light off)
+10° to +50°C (light on)
Amperage 6.8A
Door Style Solid
Insulation CFC-free, foamed polyurethane
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Model Price Quantity    

PR505755L

 
thermo scientific™
PR505755L
120V 60Hz (6.8A), NEMA 5-15P plug Plant growth unit with light CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Capacity 17.8 cu. ft. (504L)
Temperature Range (Metric) -10° to +50°C (light off)
+10° to +50°C (light on)
Amperage 6.8A
Door Style Solid
Insulation CFC-free, foamed polyurethane
Dimensions (L x W x H) Exterior 31 x 34 x 77 in. (78.7 x 86.4 x 195.6cm)
Dimensions (D x W x H) Interior 20 x 26.5 x 57 in. (50.8 x 67.3 x 144.8cm)

Utilize Thermo Scientific™ Precision™ Plant Growth Chambers with dual-lamp fluorescent lighting for uniform illumination—offering programmable temperature settings which enable users to take control of light cycles which allow for a 7-day program with 2 light cycles per day. Thermo Scientific Precision Plant Growth Chambers are best used for seed germination and plant growth applications.

  • Programmable temperature and light cycles: 7-day program with 2 light cycles per day
  • Dual-lamp fluorescent lighting provides 342-foot candles (3678 Lux) for uniform illumination
  • Long life light, rated for 24,000 hours
  • Warranty: 1 year (parts and labor)
  • Certifications: 120V unit is cCSAus listed

1 year parts and labor