Thermo Scientific™ Precision™ High-Performance Ovens
Benefit from the uniform heating, precise temperature control and fast drying of these high-performance ovens.
Specifications
Hertz 50/60Hz
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Includes Model Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Includes Model Capacity (Metric) Price Quantity  
13-248A Thermo Scientific™
6050
120V 50/60Hz Standard model 605 39.6L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-248B Thermo Scientific™
51221139
208 to 230V 50/60Hz Standard model 625 113.3L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-248C Thermo Scientific™
6056
208 to 230V 50/60Hz Standard model 645 269L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Conduct drying applications requiring high temperatures, ultra-precise temperature control and a very broad temperature range of ambient +15° to 325°C stability with Thermo Scientific™ Precision™ High-Performance Ovens. These ovens feature high-quality construction and high-efficiency insulation, providing temperature uniformity as precise as ±0.5°C with excellent stability and air exchange within the chamber.
Advanced Microprocessor PID Controls
  • Bright LED display of actual and setpoint temperatures
  • Built-in 12-hour mechanical timer along with user-adjustable safety thermostat allows for power cut-out in case of overtemperature
  • Highly responsive RTD sensor provides temperature sensitivity of ±0.1°C or better
  • High efficiency insulation ensures uniformity ±0.5° at 100°C
Energy- and Safety-Efficient Construction
  • Turbo blowers for efficient drying are side-mounted within stainless-steel chamber
  • All stainless-steel interior
  • Stainless-steel exterior door with fiberglass gasket for secure, tight seal
  • Cold-rolled, chemical-resistant steel cabinet with powder-coated finish
  • High-limit safety with built-in circuit breaker protects oven from power surges
  • Fiberglass door gasket prevents heat loss
  • Low-density heating elements ensure long life
  • Nontip shelves spaced 4cm (1.6in.) apart quickly, easily remove for thorough cleaning
Model 605P:
  • Extra baffle provides low air flow for precise drying and testing of materials with minimal disruption of volatile components
Model 625S:
  • Rear blowout panel with retaining cage and positive action front-door catches for volatile material testing
Includes:
Two nickel-plated nontip shelves
Recommended for:
  • Drug metabolism studies
  • Dry content analysis
  • Moisture determinations
  • Tobacco industry
Note:
Requires hardwired installation by a qualified technician
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
50/60Hz