Please login to your online account to display your discounted pricing
Precision™ General Purpose Water Baths Promotion
Support a wide range of applications with general purpose water baths, designed to maintain temperature from ambient to 100°C. High-performance baths range from 2L to 28L.
$1,648.02 - $1,857.09
Products
Catalog Number Mfr. No. Description Electrical Requirements Model Bath Volume Price Quantity    
TSGP05PMO
thermo scientific™
TSGP05PMO
5L General Purpose Water Bath Global Voltage 100-115V/200-230V, 50/60Hz Precision GP 05 5L (1.3 GAL) Each for $1,857.09
TSGP2SPMO
thermo scientific™
TSGP2SPMO
2L Shallow General Purpose Water Bath Global Voltage 100-115V/200-230V, 50/60Hz Precision GP 2S 2L (0.5 GAL) Each for $1,648.02
Description & Specifications
Support a wide range of applications with Thermo Scientific™ Precision™ general purpose water baths, designed to maintain water temperature from ambient to 100°C. These rugged, high-performance baths range from 2L to 28L, including shallow models. Over-temperature safety circuitry is designed to prevent thermal runaway, while new auto-on and auto-off timers allow you to optimize operation schedules. Benefit from outstanding chemical and corrosion resistance with epoxy powder-coated exterior, easily clean the chamber with its seamless stainless steel interior.
  • Free up valuable bench space with smaller footprint, compared to previous models
  • Advanced microprocessor controller is designed for extended functionality 
  • Protect your work with audible alarms
  • Conveniently save commonly used settings with four temperature presets
  • Simple to operate and monitor with icon-based graphical display
  • Enhanced hinge design supports open lid configuration and easy lid removal
  • Help prevent bath damage and overheating with low-fluid protection
  • Simplify operation with quick disconnect drain on 10, 20, and 28L and dual models