Please login to your online account to display your discounted pricing

Thermo Scientific™ Precise™ 10 x 8cm Cassette Tris-Glycine Gels

Separate proteins with outstanding resolution using these long shelf-life, short run-time, Tris-glycine gels (10 x 8cm) for SDS-PAGE.

$282.45 - $282.45

Specifications

Quantity 10 Gels
View More Specs
Products
Catalog Number Mfr. No. Formulation Sufficient For Price Quantity    

PI25261

 
thermo scientific™
25261
Precast 10% polyacrylamide gels for SDS-PAGE 10 samples per gel, 50uL capacity per well Each for $282.45

PI25265

 
thermo scientific™
25265*
Precast 8% polyacrylamide gels for SDS-PAGE 12 samples per gel, 30uL capacity per well Each for $282.45

PI25269

 
thermo scientific™
25269*
Precast 4-20% gradient polyacrylamide gels for SDS-PAGE 12 samples per gel, 30uL capacity per well Each for $282.45

PI25270

 
thermo scientific™
25270*
Precast 8% polyacrylamide gels for SDS-PAGE 15 samples per gel, 25uL capacity per well Each for $282.45

PI25271

 
thermo scientific™
25271*
Precast 10% polyacrylamide gels for SDS-PAGE 15 samples per gel, 25uL capacity per well Each for $282.45

PI25272

 
thermo scientific™
25272*
Precast 12% polyacrylamide gels for SDS-PAGE 15 samples per gel, 25uL capacity per well Each for $282.45

PI25273

 
thermo scientific™
25273*
Precast 8-16% gradient polyacrylamide gels for SDS-PAGE 15 samples per gel, 25uL capacity per well Each for $282.45
Description & Specifications

Specifications

Quantity 10 Gels

Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 × 8cm plastic cassettes to fit in Mini-PROTEAN™ gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 × 8cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 8cm cassettes fit Bio-Rad™ Mini-PROTEAN™ and Hoefer™ Mighty Small II gel tanks, as well as other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm x 8.0cm x 5mm

  • Gel Dimensions: 8cm x 6.8cm x 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank Compatibility (10cm x 8cm x 5mm): Bio-Rad Mini-PROTEAN II and 3; Bio-Rad Mini-PROTEAN Tetra Cell; Hoefer Tall Mighty Small (SE 280); Hoefer Mighty Small II (SE 260); IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 10cm Gels