Please login to your online account to display your discounted pricing

Thermo Scientific™ Precise™ 10 x 8cm Cassette Tris-Glycine Gels


Separate proteins with outstanding resolution using these long shelf-life, short run-time, Tris-glycine gels (10 x 8cm) for SDS-PAGE.

Manufacturer: thermo scientific™  25261

 View more versions of this product

Catalog No. PI25261

  • / Each
 

Description & Specifications

Specifications

Quantity 10 Gels
Formulation Precast 10% polyacrylamide gels for SDS-PAGE
Description 10% Tris-Glycine Gels, 10 x 8cm, 10-well
Sufficient For 10 samples per gel, 50uL capacity per well

Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 × 8cm plastic cassettes to fit in Mini-PROTEAN™ gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 × 8cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 8cm cassettes fit Bio-Rad™ Mini-PROTEAN™ and Hoefer™ Mighty Small II gel tanks, as well as other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm x 8.0cm x 5mm

  • Gel Dimensions: 8cm x 6.8cm x 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank Compatibility (10cm x 8cm x 5mm): Bio-Rad Mini-PROTEAN II and 3; Bio-Rad Mini-PROTEAN Tetra Cell; Hoefer Tall Mighty Small (SE 280); Hoefer Mighty Small II (SE 260); IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 10cm Gels