missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Precise™ 10 x 10cm Cassette Tris-Glycine Gels

Separate proteins with outstanding resolution using these long shelf-life, short run-time, Tris-glycine gels (10 x 10cm) for SDS-PAGE.

Manufacturer:  Thermo Scientific™ 25253*

 View more versions of this product

Catalog No. PI25253



Description

Description

These gels have been discontinued and are replaced by the Novex WedgeWell line.
Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 x 10cm plastic cassettes to fit in Novex™ XCell gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 x 10cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well and 12-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 60-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 10cm cassettes fit the Novex™ XCell and a other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm × 10cm × 7mm

  • Gel Dimensions: 8cm × 8.8cm × 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank CompatibilityCompatibility (10cm x 10cm x 7mm): Novex XCell I and II; Novex XCell II™ Surelock™; IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 8cm Gels

Specifications

Specifications

8-16% Tris-Glycine Gels, 10 x 10cm, 12-well
Precast 8-16% gradient polyacrylamide gels for SDS-PAGE
10 Gels
12 samples per gel, 30uL capacity per well
Documents