Please login to your online account to display your discounted pricing

Alfa Aesar™ Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL

Chemical Identifiers

Chemical Name or Material Precious Metals, plasma standard solution
Name Note Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL
Purity Grade Specpure™
MDL Number MFCD00151264
UN Number UN1789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14652AE

  SDS
alfa aesar™
14652AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14652AP

  SDS
alfa aesar™
14652AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Precious Metals, plasma standard solution
Quantity 100mL
TSCA TSCA
Name Note Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL
Purity Grade Specpure™
Physical Form Liquid
Concentration Matrix: 20% HCl
MDL Number MFCD00151264
UN Number UN1789
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.