missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014652AP
Description
Precious Metals, plasma standard solution is useful as a standard for the determination of metals by inductively coupled plasma-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Precious Metals, plasma standard solution | |
Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL | |
MFCD00151264 | |
Specpure™ | |
Liquid |
Matrix: 20% HCl | |
500 mL | |
UN1789 | |
Miscible with water |
RUO – Research Use Only