Learn More
Praziquantel, 98%, Thermo Scientific Chemicals
Used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2 channels and is a potential small molecule neurogenic.
Supplier: Thermo Scientific Chemicals J6618806
Description
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPraziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.
Solubility
Freely soluble in ethanol or dichloromethane. Slightly soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Praziquantel | |
136°C to 138°C | |
1377°C | |
C19H24N2O2 | |
5 g | |
14,7716 | |
Freely soluble in ethanol or dichloromethane; Slightly soluble in water | |
C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2 | |
312.413 | |
312.41 | |
Powder |
55268-74-1 | |
White | |
>110°C (230°F) | |
MFCD00058531 | |
761557 | |
praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum | |
FSVJFNAIGNNGKK-UHFFFAOYSA-N | |
2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one | |
4891 | |
98% |
Chemical Identifiers
55268-74-1 | |
312.413 | |
FSVJFNAIGNNGKK-UHFFFAOYSA-N | |
4891 | |
C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2 |
C19H24N2O2 | |
MFCD00058531 | |
praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum | |
2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one |
Safety and Handling
EINECSNumber : 259-559-6
RTECSNumber : UQ4150000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only