Please login to your online account to display your discounted pricing

Praseodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLPR22Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLPR2-2Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 1 Causes skin irritation, serious eye irritation. May cause respiratory irritation
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Quantity 125mL
DOT Information Hazardous
Recommended Storage Ambient
Solvent or Matrix 2% HNO3
Purity Grade Plasma Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLPR2-2Y, -2X    10,000    PLPR2-3Y, -3X
CAS Max % 2.0
CAS Min % 97.9
CAS 7697-37-2
CAS 7440-10-0
CAS Min % 0.1
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 2.0
CAS Max % 97.9
Chemical Name or Material Praseodymium Standard

Specifications

Chemical Name or Material Praseodymium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation. May cause respiratory irritation

Ambient