Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium rod, 12.7mm (0.5 in.) dia., 99.5% (metals basis excluding Ta)

Chemical Identifiers

Chemical Name or Material Praseodymium rod
CAS 7440-10-0
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00011174
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40296DH

 
alfa aesar™
40296DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40296DJ

 
alfa aesar™
40296DJ
50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40296DM

 
alfa aesar™
40296DM
100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40296HF

 
alfa aesar™
40296HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cast Rod, dia. is approximate
Quantity 25mm
Packaging Mylar under Argon
Chemical Name or Material Praseodymium rod
Odor Odorless
TSCA TSCA
Name Note 12.7mm (0.5 in.) dia.
Assay 99.5%
Assay Percent Notes (metals basis excluding Ta)
MDL Number MFCD00011174
CAS 7440-10-0
Storage Note 1
  • Packaged under argon
Weight ≈0.86g/mm

  • Packaged under argon

Safety and Handling

  • Packaged under argon