Learn More
Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04591014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Silver | |
7440-10-0 | |
Pr | |
UN3089 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
−200 Mesh | |
Air and moisture sensitive |
Praseodymium powder | |
(REO) | |
MFCD00011174 | |
25 g | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
99.9% | |
Odorless | |
Powder |
Chemical Identifiers
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Safety and Handling
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber : 231-120-3
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only