Learn More
Alfa Aesar™ Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO)
Manufacturer: Alfa Aesar™ 1318014
Specifications
Praseodymium(III) sulfate octahydrate | |
(REO) | |
MFCD00149832 | |
praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3] | |
714.103 | |
714.12 (570.00 Anhydrous) | |
99.99% | |
Odorless |
13510-41-3 | |
H16O20Pr2S3 | |
14,7712 | |
NIJXLQIGVGZNKP-UHFFFAOYSA-H | |
praseodymium(3+);trisulfate;octahydrate | |
25022097 | |
Powder | |
Reacton™ | |
25g |
Chemical Identifiers
13510-41-3 | |
714.103 | |
NIJXLQIGVGZNKP-UHFFFAOYSA-H | |
25022097 | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3] |
H16O20Pr2S3 | |
MFCD00149832 | |
praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate | |
praseodymium(3+);trisulfate;octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA