Please login to your online account to display your discounted pricing

Praseodymium(III) sulfate octahydrate, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Praseodymium(III) sulfate octahydrate
Name Note Rare Earth Oxides Content
CAS 13510-41-3
Physical Form Powder
Assay Percent Range 99.9% (REO)
Molecular Formula Pr2(SO4)3·8H2O
Formula Weight 714.12/mol; 570.00 (anhydrous)
MDL Number MFCD00149832
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1460714

  SDS
alfa aesar
1460714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1460722

  SDS
alfa aesar
1460722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 714.12/mol; 570.00 (anhydrous)
Physical Form Powder
Quantity 25g
Color Green
Anhydrous or Hydrate Octahydrate
EINECS Number 233-622-8
Density 2.827g/cm3
Assay Percent Range 99.9% (REO)
Appearance Green powder
CAS 13510-41-3
Odor Odorless
Name Note Rare Earth Oxides Content
Chemical Name or Material Praseodymium(III) sulfate octahydrate
MDL Number MFCD00149832
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Molecular Formula Pr2(SO4)3·8H2O
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.