missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium(III) sulfate octahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1460722

 View more versions of this product

Catalog No. AA1460722



Specifications

Specifications

Praseodymium(III) sulfate octahydrate
13510-41-3
H16O20Pr2S3
Green powder
NIJXLQIGVGZNKP-UHFFFAOYSA-H
praseodymium(3+);trisulfate;octahydrate
25022097
Powder
Octahydrate
100g
Rare Earth Oxides Content
99.9% (REO)
MFCD00149832
praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3]
714.103
714.12 (570.00 Anhydrous)
Odorless
Green

Chemical Identifiers

13510-41-3
714.103
NIJXLQIGVGZNKP-UHFFFAOYSA-H
25022097
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3]
H16O20Pr2S3
MFCD00149832
praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate
praseodymium(3+);trisulfate;octahydrate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 233-622-8

SDS Documents