Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Chemical Identifiers

Chemical Name or Material Praseodymium(III) perchlorate
CAS 13498-07-2
Purity Grade Reagent Grade
Formula Weight 439.26
MDL Number MFCD00243288
UN Number UN3211
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40569NC

  SDS
alfa aesar™
40569NC
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40569NE

  SDS
alfa aesar™
40569NE
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 439.26
Quantity 10g
CAS 13498-07-2
UN Number UN3211
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 50% w/w aq. soln.
Purity Grade Reagent Grade
Physical Form Liquid
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
Chemical Name or Material Praseodymium(III) perchlorate
Odor Odorless
MDL Number MFCD00243288
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.