Please login to your online account to display your discounted pricing

Praseodymium(III) nitrate hydrate, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Praseodymium(III) nitrate hydrate
Name Note Rare Earth Oxides Content
Physical Form Crystalline
Assay Percent Range 99.9%
Molecular Formula Pr(NO3)3·xH2O
MDL Number MFCD00011175
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1290909

  SDS
alfa aesar
1290909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1290918

  SDS
alfa aesar
1290918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1290930

  SDS
alfa aesar
1290930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12909A1

  SDS
alfa aesar
12909A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 326.92 (Anhydrous)
Quantity 10g
Color Green
DOT Information DOT Class: 5.1, Packaging Group: III
EINECS Number 233-796-5
Name Note Rare Earth Oxides Content
Anhydrous or Hydrate Hydrate
Physical Form Crystalline
Molecular Formula Pr(NO3)3·xH2O
Appearance Green crystals
Odor Odorless
Assay Percent Range 99.9%
Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Chemical Name or Material Praseodymium(III) nitrate hydrate
MDL Number MFCD00011175
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.