missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium(III) nitrate hydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1290930

 View more versions of this product

Catalog No. AA1290930



Specifications

Specifications

Praseodymium(III) nitrate hydrate
15878-77-0
H12N3O15Pr
Green crystals
LXXCECZPOWZKLC-UHFFFAOYSA-N
praseodymium(3+);trinitrate;hexahydrate
204170
Crystalline
Hydrate
250g
Rare Earth Oxides Content
99.9%
MFCD00011175
praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Pr+3]
435.01
326.92 (anhydrous)
Odorless
Green

Chemical Identifiers

15878-77-0
435.01
LXXCECZPOWZKLC-UHFFFAOYSA-N
204170
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Pr+3]
H12N3O15Pr
MFCD00011175
praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
praseodymium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-796-5

Documents