Learn More
Praseodymium(III, IV) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01091406
Description
It is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Praseodymium(III, IV) oxide | |
12037-29-5 | |
5 g | |
O11Pr6 | |
Insoluble in water; moderately soluble in strong mineral acids. | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] | |
1021.44 | |
99.996% |
2500°C | |
Powder | |
(REO) | |
MFCD00011178 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
hexapraseodymium(3+) undecaoxidandiide | |
1021.43 | |
Reacton™ |
Chemical Identifiers
12037-29-5 | |
1021.44 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] |
O11Pr6 | |
MFCD00011178 | |
hexapraseodymium(3+) undecaoxidandiide |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 234-857-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only