missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III, IV) oxide, REacton™, 99.996% (REO)

Manufacturer:  Alfa Aesar™ 1091414

 View more versions of this product

Catalog No. AA1091414



Specifications

Specifications

Praseodymium(III, IV) oxide
(REO)
1021.43
99.996%
2500°C
12037-29-5
MFCD00011178
Powder
Reacton™
25g

Chemical Identifiers

12037-29-5
MFCD00011178
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents