missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III, IV) oxide, 99% (REO)

Manufacturer:  Alfa Aesar™ 4459430

 View more versions of this product

Catalog No. AA4459430



Specifications

Specifications

Praseodymium (III, IV) oxide
(REO)
MFCD00011178
1021.43
99%
250g
12037-29-5
Pr6O11
Insoluble in water and only soluble in strong acid.
Powder
2500°C

Chemical Identifiers

12037-29-5
Pr6O11
MFCD00011178
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 234-857-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents