Learn More
Praseodymium(III,IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 194620500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Praseodymium(III, IV) oxide | |
12037-29-5 | |
Powder | |
99.90% | |
Pr6O11 | |
14, 7712 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
hexapraseodymium(3+) undecaoxidandiide | |
1021.44 | |
Trace Metal Basis |
99.9% | |
Black or Brown to Green | |
50 g | |
O11Pr6 | |
MFCD00011178 | |
Solubility in water: insoluble | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] | |
1021.44 | |
99.9% (trace metals basis) | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face,hands and any exposed skin thoroug
GHS Signal Word: Warning
EINECSNumber : 234-857-9
RTECSNumber : TU1480000
TSCA : TSCA
RUO – Research Use Only