Please login to your online account to display your discounted pricing

Praseodymium(III,IV) oxide, 99.9%, (trace metal basis), ACROS Organics™

$151.46 - $151.46

Chemical Identifiers

Chemical Name or Material Praseodymium(III, IV) oxide
Name Note 99.9%
CAS 12037-29-5
Purity Grade For trace metal basis analysis
Assay Percent Range 99.90%
Molecular Formula O11Pr6
Linear Formula Pr6O11
Formula Weight 1021.44
MDL Number MFCD00011178
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194620500

  SDS
thermo fisher scientific
194620500
Glass bottle 50g Each for $151.46
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 1021.44
Quantity 50g
Color Dark green to dark brown or black
RTECS Number TU1480000
EINECS Number 234-857-9
TSCA TSCA
Purity Grade For trace metal basis analysis
Merck Index 14, 7712
Packaging Glass bottle
Total Impurity 0.1% max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material Praseodymium(III, IV) oxide
MDL Number MFCD00011178
Linear Formula Pr6O11
CAS 12037-29-5
Assay Percent Range 99.90%
Molecular Formula O11Pr6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99.9%
Health Hazard 1 HYGROSCOPIC

for ultra trace analysis

Safety and Handling

HYGROSCOPIC

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.