missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium(III, IV) oxide, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03627618

 View more versions of this product

Catalog No. AA3627618



Description

Description

Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Praseodymium(III, IV) oxide
12037-29-5
50 g
O11Pr6
Insoluble in water. Moderately soluble in strong mineral acids.
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
1021.44
99.9%
2500°C
Sintered Lump
(REO)
MFCD00011178
HPZIIFVSYNLWNX-UHFFFAOYSA-N
hexapraseodymium(3+) undecaoxidandiide
1021.43

Chemical Identifiers

12037-29-5
1021.44
HPZIIFVSYNLWNX-UHFFFAOYSA-N
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
O11Pr6
MFCD00011178
hexapraseodymium(3+) undecaoxidandiide
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 234-857-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only