Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III, IV) oxide, 99.5% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III, IV) oxide
CAS 12037-29-5
Formula Weight 1021.43
MDL Number MFCD00011178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1123414

  SDS
alfa aesar™
1123414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1123422

  SDS
alfa aesar™
1123422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11234A1

  SDS
alfa aesar™
11234A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1021.43
Chemical Name or Material Praseodymium(III, IV) oxide
TSCA TSCA
Physical Form Powder
Assay 99.5%
Assay Percent Notes (REO)
MDL Number MFCD00011178
CAS 12037-29-5
Melting Point 2500°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.