Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) iodide, ultra dry, 99.9+% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) iodide
CAS 13813-23-5
Name Note Ultra dry
Linear Formula PrI3
Formula Weight 521.62
MDL Number MFCD00049513
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3938503

  SDS
alfa aesar™
3938503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3938506

  SDS
alfa aesar™
3938506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 521.62
Packaging Ampule
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Praseodymium(III) iodide
Melting Point 737°C
Assay Percent Notes (REO)
Assay ≥99.9%
TSCA TSCA
Storage Note 1
  • Ampouled under argon
CAS 13813-23-5
Physical Form Crystalline/Needles
MDL Number MFCD00049513
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Quantity 1g
Sensitivity Light Sensitive & Hygroscopic
Linear Formula PrI3
Health Hazard 1 Danger
Name Note Ultra dry

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

  • Ampouled under argon