missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) fluoride, anhydrous, REacton™, 99.9895% (REO)

Manufacturer:  Alfa Aesar™ 1124104

 View more versions of this product

Catalog No. AA1124104



Specifications

Specifications

Praseodymium(III) fluoride
13709-46-1
PrF3
UN3288
Powder
Reacton™
Odorless
Green
2g
Anhydrous
(REO)
MFCD00016287
197.9
99.9895%
6.2g/mL
Hygroscopic
1395°C

Chemical Identifiers

13709-46-1
MFCD00016287
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents