Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) fluoride
CAS 13709-46-1
Name Note Anhydrous
Purity Grade Reacton™
Formula Weight 197.9
MDL Number MFCD00016287
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1365814

  SDS
alfa aesar™
1365814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1365822

  SDS
alfa aesar™
1365822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.9
UN Number UN3288
Purity Grade Reacton™
Chemical Name or Material Praseodymium(III) fluoride
TSCA TSCA
Melting Point 1395°C
Assay 99.9%
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Color Green
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Anhydrous
Physical Form Crystalline
Quantity 25g
Assay Percent Notes (REO)
Odor Odorless
MDL Number MFCD00016287
Density 6.2g/mL
Health Hazard 1 Danger
Sensitivity Hygroscopic
CAS 13709-46-1
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.