Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) chloride, ultra dry, 99.99% (REO)

$680.36 - $680.36

Chemical Identifiers

Chemical Name or Material Praseodymium(III) chloride
CAS 10361-79-2
Name Note Ultra dry
Formula Weight 247.27
MDL Number MFCD00011176
Merck Index 14,7712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3938614

 
alfa aesar™
3938614
25g Each for $680.36

AA3938606

 
alfa aesar™
3938606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.27
Packaging Ampule
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Praseodymium(III) chloride
TSCA TSCA
Odor Odorless
Name Note Ultra dry
Physical Form Powder
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00011176
CAS 10361-79-2
Merck Index 14,7712
Boiling Point 1700°C
Melting Point 786°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Storage Note 1
  • Ampouled under argon
Density 4.02g/mL

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

  • Ampouled under argon