Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) chloride, ultra dry, 99.95% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) chloride
CAS 10361-79-2
Name Note Ultra dry
Linear Formula PrCl3
Formula Weight 247.27
MDL Number MFCD00011176
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3781503

  SDS
alfa aesar™
H3781503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3781506

  SDS
alfa aesar™
H3781506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3781514

  SDS
alfa aesar™
H3781514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.27
Packaging Ampule
Chemical Name or Material Praseodymium(III) chloride
TSCA TSCA
Odor Odorless
Name Note Ultra dry
Physical Form Powder
Assay 99.95%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula PrCl3
MDL Number MFCD00011176
CAS 10361-79-2
Boiling Point 1700°C
Melting Point 786°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Storage Note 1
  • Ampouled under argon
Density 4.02g/mL
RTECS Number TU0175000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

  • Ampouled under argon