missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) chloride, ultra dry, 99.95% (REO)

Manufacturer:  Alfa Aesar™ H3781514

 View more versions of this product

Catalog No. AAH3781514


 

Description

Description

  • Ampouled under argon
Specifications

Specifications

Praseodymium(III) chloride
10361-79-2
(REO)
MFCD00011176
Powder
Odorless
Hygroscopic
786°C
Ultra dry
99.95%
PrCl3
247.27
4.02g/mL
Ampule
1700°C
25g

Chemical Identifiers

10361-79-2
MFCD00011176
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TU0175000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents