Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) chloride hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) chloride hydrate
CAS 19423-77-9
Purity Grade Reacton™
Formula Weight 247.27 (Anhydrous)
MDL Number MFCD00149828
Merck Index 14,7712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1123814

  SDS
alfa aesar™
1123814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1123822

  SDS
alfa aesar™
1123822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.27 (Anhydrous)
Assay Percent Notes (REO)
Purity Grade Reacton™
Quantity 25g
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Assay 99.99%
TSCA TSCA
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Praseodymium(III) chloride hydrate
Physical Form Wet Crystalline Aggregates
Merck Index 14,7712
Health Hazard 1 Warning
Melting Point 115°C
MDL Number MFCD00149828
RTECS Number TU0175000
Odor Odorless
CAS 19423-77-9
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.