Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) carbonate octahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) carbonate octahydrate
CAS 14948-62-0
Purity Grade Reacton™
Formula Weight 605.97 (461.85 Anhydrous)
MDL Number MFCD00211307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1318206

  SDS
alfa aesar™
1318206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1318214

  SDS
alfa aesar™
1318214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1318222

  SDS
alfa aesar™
1318222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 605.97 (461.85 Anhydrous)
TSCA TSCA
MDL Number MFCD00211307
Odor Odorless
Chemical Name or Material Praseodymium(III) carbonate octahydrate
Physical Form Crystalline
Assay Percent Notes (REO)
CAS 14948-62-0
Assay 99.99%
Purity Grade Reacton™
Quantity 5g