missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) carbonate octahydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1318214

 View more versions of this product

Catalog No. AA1318214



Specifications

Specifications

Praseodymium(III) carbonate octahydrate
(REO)
MFCD00211307
SEHQEMTYQGRHIC-UHFFFAOYSA-H
praseodymium(3+);tricarbonate;octahydrate
25022096
Crystalline
Reacton™
25g
14948-62-0
C3H16O17Pr2
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]
605.959
605.97 (461.85 Anhydrous)
99.99%
Odorless

Chemical Identifiers

14948-62-0
605.959
SEHQEMTYQGRHIC-UHFFFAOYSA-H
25022096
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]
C3H16O17Pr2
MFCD00211307
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
praseodymium(3+);tricarbonate;octahydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents