Please login to your online account to display your discounted pricing

Praseodymium(III) acetylacetonate hydrate, +99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Praseodymium(III) acetylacetonate hydrate
Assay Percent Range 99+%
Molecular Formula C15H21O6Pr·xH2O
Linear Formula Pr[OCH(CH3)CH2C(O)CH3]3
Formula Weight 438.24
MDL Number MFCD08272322
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381460010

  SDS
thermo fisher scientific
AC381460010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 438.24
Quantity 1g
Color Off-white to pale green
Praseodymium (Pr) 99.0 min.
Assay Percent Range 99+%
Linear Formula Pr[OCH(CH3)CH2C(O)CH3]3
MDL Number MFCD08272322
Chemical Name or Material Praseodymium(III) acetylacetonate hydrate
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C15H21O6Pr·xH2O
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.