Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium(III) acetate hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Praseodymium(III) acetate hydrate
CAS 6192-12-7
Formula Weight 318.03 (Anhydrous)
MDL Number MFCD00150121
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1461114

  SDS
alfa aesar™
1461114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1461122

  SDS
alfa aesar™
1461122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.03 (Anhydrous)
Chemical Name or Material Praseodymium(III) acetate hydrate
TSCA TSCA
Physical Form Crystalline Aggregates
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00150121
CAS 6192-12-7
Quantity 25g