missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium foil, 1.0mm (0.04 in.) thick, 99.5% (REO)

Manufacturer:  Alfa Aesar™ 13963FF

 View more versions of this product

Catalog No. AA13963FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Praseodymium foil
White
1.0mm (0.04 in.) thick
Odorless
(REO)
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
Foil
≈4.23g/25x25mm
25 x 25mm
7440-10-0
Mylar under Argon
Pr
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
[Pr]
140.908
CHEBI:49828
99.5%

Chemical Identifiers

7440-10-0
140.908
PUDIUYLPXJFUGB-UHFFFAOYSA-N
23942
praseodymium
Pr
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
CHEBI:49828
[Pr]
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents