missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010305FF

 View more versions of this product

Catalog No. AA10305FF



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gray
0.25mm (0.01 in.) thick
(REO)
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
[Pr]
140.91
CHEBI:49828
Odorless
≈1.27g/25x25mm
Praseodymium foil
7440-10-0
Pr
25 x 25 mm
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
99.5%
Mylar under Argon
Foil

Chemical Identifiers

7440-10-0
140.91
PUDIUYLPXJFUGB-UHFFFAOYSA-N
23942
praseodymium
Pr
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
CHEBI:49828
[Pr]
Safety and Handling

Safety and Handling

EINECSNumber : 231-120-3

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only