missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
$811.22 - $811.22
Chemical Identifiers
CAS | 7440-10-0 |
---|---|
Molecular Formula | Pr |
Molecular Weight (g/mol) | 140.91 |
MDL Number | MFCD00011174 |
InChI Key | PUDIUYLPXJFUGB-UHFFFAOYSA-N |
Synonym | unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg |
PubChem CID | 23942 |
ChEBI | CHEBI:49828 |
IUPAC Name | praseodymium |
SMILES | [Pr] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA10305FF
|
Thermo Scientific Chemicals
010305FF |
25 x 25 mm |
Each for $811.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Specifications
Gray | |
Pr | |
25 x 25 mm | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
99.5% | |
Mylar under Argon | |
Foil | |
(REO) |
7440-10-0 | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
Odorless | |
≈1.27g/25x25mm | |
Praseodymium foil, 0.25mm (0.01 in.) thick |
Safety and Handling
EINECSNumber : 231-120-3
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only