missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Praseodymium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 13962FF

 View more versions of this product

Catalog No. AA13962FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Praseodymium foil
25 x 25mm
7440-10-0
Mylar under Argon
Pr
UN3178
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
Foil
≈0.42g/25x25mm
0.1mm (0.004 in.) thick
Odorless
(REO)
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
[Pr]
140.908
CHEBI:49828
99.9%

Chemical Identifiers

7440-10-0
140.908
PUDIUYLPXJFUGB-UHFFFAOYSA-N
23942
praseodymium
Pr
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
CHEBI:49828
[Pr]
Safety and Handling

Safety and Handling

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents