missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pranlukast, TCI America™

Manufacturer:  TCI America P2115100MG

 View more versions of this product

Catalog No. P2115100MG



Specifications

Specifications

Pranlukast
≥98.0% (HPLC)
MFCD00864631
C1=CC=C(C=C1)CCCCOC2=CC=C(C=C2)C(=O)NC3=CC=CC4=C3OC(=CC4=O)C5=NNN=N5
481.512
481.51
White-Yellow
100mg
103177-37-3
C27H23N5O4
NBQKINXMPLXUET-UHFFFAOYSA-N
N-[4-oxo-2-(2H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
4887
Crystal-Powder at 20°C
233°C

Chemical Identifiers

103177-37-3
481.512
NBQKINXMPLXUET-UHFFFAOYSA-N
N-[4-oxo-2-(2H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
C27H23N5O4
MFCD00864631
4887
C1=CC=C(C=C1)CCCCOC2=CC=C(C=C2)C(=O)NC3=CC=CC4=C3OC(=CC4=O)C5=NNN=N5
Safety and Handling

Safety and Handling

RTECSNumber : CV5647500

TSCA : No

Recommended Storage : Refrigerator

Documents