Please login to your online account to display your discounted pricing

Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

$308.98 - $1,421.29

Specifications

Antigen PR3 (Proteinase 3) Ab-1
Clone MCPR3-2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1333P

 
thermo scientific™
MS1333P
200μg/mL 1mL Each for $1,292.63

MS1333P0

 
thermo scientific™
MS1333P0
200μg/mL 100μL Each for $308.98

MS1333P1

 
thermo scientific™
MS1333P1
200μg/mL 500μL Each for $766.37

MS1333P1ABX

 
thermo scientific™
MS1333P1ABX
1mg/mL 100μL Each for $845.90

MS1333PABX

 
thermo scientific™
MS1333PABX
1mg/mL 200μL Each for $1,421.29
Description & Specifications

Specifications

Antigen PR3 (Proteinase 3) Ab-1
Clone MCPR3-2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen HMC-1/PR3 cell granule extract
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Proteinase 3 (PR3), a constituent of azurophil granules of neutrophils (polymorphonuclear cells, PMNs), is the target antigen for most anti-neutrophil cytoplasmic antibodies (c-ANCA) in Wegener's granulomatosis (WG).

Host Species: Mouse

Clone: MCPR3-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HMC-1/PR3 cell granule extract

Molecular Weight: 32-40kDa

Positive Control: HMC-1/PR3-S176 cells

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO