Please login to your online account to display your discounted pricing

Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS1333PABX

 View more versions of this product

Catalog No. MS1333PABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a kappa
Cross Reactivity Human
Host Species Mouse
Quantity 200µL
Format Concentrated
Immunogen HMC-1/PR3 cell granule extract
Clone MCPR3-2
Regulatory Status RUO
Applications Immunofluorescence
Antigen PR3 (Proteinase 3) Ab-1
Research Discipline Proteases And Inhibitors
Conjugate Unlabeled
Concentration 1mg/mL
Type Primary
Classification Monoclonal

Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Proteinase 3 (PR3), a constituent of azurophil granules of neutrophils (polymorphonuclear cells, PMNs), is the target antigen for most anti-neutrophil cytoplasmic antibodies (c-ANCA) in Wegener's granulomatosis (WG).

Host Species: Mouse

Clone: MCPR3-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HMC-1/PR3 cell granule extract

Molecular Weight: 32-40kDa

Positive Control: HMC-1/PR3-S176 cells

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO