Please login to your online account to display your discounted pricing

Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS1333P1ABX

 View more versions of this product

Catalog No. MS1333P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen PR3 (Proteinase 3) Ab-1
Applications Immunofluorescence
Classification Monoclonal
Clone MCPR3-2
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen HMC-1/PR3 cell granule extract
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ PR3 (Proteinase 3) Ab-1, Mouse Monoclonal Antibody.

Proteinase 3 (PR3), a constituent of azurophil granules of neutrophils (polymorphonuclear cells, PMNs), is the target antigen for most anti-neutrophil cytoplasmic antibodies (c-ANCA) in Wegener's granulomatosis (WG).

Host Species: Mouse

Clone: MCPR3-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HMC-1/PR3 cell granule extract

Molecular Weight: 32-40kDa

Positive Control: HMC-1/PR3-S176 cells

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO