Please login to your online account to display your discounted pricing

PPT, Tocris

Subtype-selective ERα agonist

$220.30 - $924.23

Chemical Identifiers

Chemical Name or Material PPT
Assay Percent Range >99%
Molecular Formula C24H22N2O3
Formula Weight 386.45
Synonym 4,4',4''-(4-Propyl-[1H]-pyrazole-1,3,5-triyl)trisphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

142610

 
r & d systems
RDS142610
10mg Each for $220.30

142650

 
r & d systems
RDS142650
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 386.45
Quantity 10mg
Chemical Name or Material PPT
Molecular Formula C24H22N2O3
CAS 263717-53-9
Synonym 4,4',4''-(4-Propyl-[1H]-pyrazole-1,3,5-triyl)trisphenol
Assay Percent Range >99%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C